Imprimer

6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (par CCM)

Marques : Enzo Life Sciences
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): 7-Nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrileCNQX6-Cyano-7-nitroquinoxaline-2,3(1H,4H)-dione

ALEXALX550042M025EA 373 EUR
ALEXALX550042M025 ALEXALX550042M005 ALEXALX550042M050 ALEXALX550042M010
6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (par CCM)
6-Cyano-7-nitroquinoxaline-2,3-dione

AMPA/kainate receptor antagonist


  • Formulation: Off-white to yellow powder.


Potent, competitive kainate/quisqualate (non-NMDA) receptor antagonist. For the more soluble form see CNQX . disodium salt (Prod. No. ALX-550-283).

Formule: C₉H₄N₄O₄
Poids moléculaire: 232,16 g/mol
Numéro MDL: MFCD00069232
Numéro CAS: 115066-14-3
https://fr.vwr.com/stibo/web/std.lang.all/07/95/45390795.jpg

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR