Imprimer

Tetramethylammonium tris(pentafluoroethyl)trifluorophosphate, Sigma-Aldrich®

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5


M-Clarity™ quality level = MQ200

4.90025.0500EA 5670 EUR
4.90025.0500
Tetramethylammonium tris(pentafluoroethyl)trifluorophosphate, Sigma-Aldrich®
Tetramethylammonium tris(pentafluoroethyl)trifluorophosphate
Formule: C₁₀H₁₂F₁₈NP
Numéro CAS: 394692-83-2

Commander maintenant

Résultats des tests de spécification

Melting Point 115 °C
Molar Mass 519.15 g/mol
pH 6 (0.01 g/ml, H2O, 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR