Imprimer
Marker pen, Write-ON™
Fournisseur: Simport
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SIMPM495-12EA 72.2 EUR
SIMPM495-12
Marker pen, Write-ON™
This pen is specially suited to be used on anterior surfaces and sides of histology cassettes.
  • Ink will dry almost instantly
  • Ink withstands temperatures up to 195 °C and is resistant to solvents