Imprimer
Cleanroom wipes, EvoControl
Fournisseur: Vileda Professional
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VILE142870EA 2100 EUR
VILE142870 VILE142854 VILE142855 VILE142856 VILE142857
Cleanroom wipes, EvoControl
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
These microfibre cloth are manufactured from endless Evolon® microfibres measuring 0,15 dtex (10 times finer than standard polyester microfibres). The unique fabric combines very good textile and mechanical properties. With one single wiping movement, 99,999% of all bacteria are removed from the surface, even without the use of disinfectants.
  • Designed to hold the right amount of liquid, distributing it readily and evenly onto the surface
  • Microfibres reach even into microscopic surface pores and thus help remove contamination thouroughly
  • High contact area of the wipe on the surface, for superior cleaning results
  • Laser cut sealed edges to avoid particle release
EvoControl 200: for ISO class 6 - 9 environments
- Disposable precision cleaning wipe for fine wiping
- Laser cut for areas where moderate contamination control is required
EvoControl 400: for ISO class 4 - 9 environments
- Disposable liquid cleaning wipe for contamination removal
- Pre-laundered, laser cut and designed for the removal of acids and chemical spills
EvoControl 500: for ISO class 4 - 9 environments
- Reusable contamination collector and residue remover
- Designed for liquid removal
- Pre-laundered, laser-cut durable: enables microporeus cleaning to improve validation SOP’s
- Can be washed up to 10 times
EvoControl 500+: for ISO class 5 - 9 environments
- Reusable contamination collector and residue remover
- Designed for liquid removal
- Pre-laundered, laser-cut durable: enables microporeus cleaning to improve validation SOP’s
- Can be washed up to 20 times
Colour: White