Imprimer
Laboratory reactor systems, LR 1000 packages
Fournisseur: IKA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IKAA8040100EA 9910 EUR
IKAA8040100 IKAA8040200 IKAA8040102 IKAA8040103 IKAA8040202 IKAA8040203
Laboratory reactor systems, LR 1000 packages
Systèmes de réacteurs de laboratoire
The LR 1000 basic is a cost-efficient, modular laboratory reactor designed for optimising chemical reaction processes as well as mixing, dispersing, and homogenisation applications at the laboratory scale.
  • Digital display for speed and temperature monitoring
  • Vacuum valve provided
  • Adjustable safety circuit
  • Integrated safety shutdown when vessel or lid is removed from base
The reactor is designed for the unique application requirements that define the cosmetic and pharmaceutical industries. The temperature of the medium can be as high as 120 °C.
The temperature of the heating source, which is located at the bottom of the vessel, is controlled accordingly to reach the set temperature inside the medium.
Informations de commande: A PT100,30 temperature sensor and the corresponding receptacle LR 1000,61 for the lid are included with this package.
Attention: * RT=Ambient
* RT = Ambient