Imprimer
Gas leak detector
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HYPE66002-001EA 1430 EUR
HYPE66002-001
Gas leak detector
This handheld instrument can detect minute leaks of laboratory gases.
  • Automatic shut-off function
  • Small probe detector to reach difficult and confined locations
  • Both visual and audible alarms: LED light to indicate the intensity of the leak
Specifically designed for gas chromatography.