Imprimer
Chemvac pump systems
Fournisseur: Gardner Denver
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ILMV109030EA 4630 EUR
ILMV109030
Chemvac pump systems
These pump systems consist of a two-stage rotary vane pump P 6 Z and a two-stage diaphragm pump MPC 101 Z. The Chemvac combination pump draws a deep, high-flow vacuum, suitable for freeze dry applications with a powerful rotary vane pump.
  • The harmful ingested fumes compromise the primary pump’s oil that are promptly degassed by Chemvac’s secondary vacuum, a corrosion resistant PTFE diaphragm pump. Both pumps are electrical and vacuum-connected. On the outtake of the rotary vane pump is a separator for the collection of condensates. The solenoid valve block is part of the delivery.