Imprimer
Centrifuge bottles, PP/PC, with screw cap, Corning®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Corning®
734-4270EA 257 EUR
734-4270 734-4271 734-4267 734-4268 734-4269 734-4264 734-4265 734-4266
Centrifuge bottles, PP/PC, with screw cap, Corning®
Flacons Flacons à centrifuger
PP, translucent or PC, clear.
  • Clear graduations and textured marking spot
  • Specially designed cap with a built-in easy grip feature
  • Caps are not interchangeable among different size/material bottles
  • Extra-thick walls for durability
  • Wide-mouth bottle design for improved liquid handling
Centrifuge bottles for large volume processing in cell harvesting, pelleting and protein purification. Allow researchers to decrease the number of centrifugation cycles, improve the laboratory efficiency and boost productivity. Two cap styles: standard and high performance with silicone O-ring. Leak-free processing at maximum rated speeds (Fill volume should not exceed 80 % of the total bottle capacity).
  • Clear graduations and textured marking spot
  • Specially designed cap with a built-in easy grip feature
  • Caps are not interchangeable among different size/material bottles
  • Extra-thick walls for durability
  • Wide-mouth bottle design for improved liquid handling
Centrifuge bottles for large volume processing in cell harvesting, pelleting and protein purification. Allow researchers to decrease the number of centrifugation cycles, improve the laboratory efficiency and boost productivity. Two cap styles: standard and high performance with silicone O-ring. Leak-free processing at maximum rated speeds (Fill volume should not exceed 80% of the total bottle capacity).