Imprimer
Colonnes capillaires, phases traditionnelles, SLB-5ms, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28469-UEA 539 EUR
28469-U 28465-U 28476-U 28467-U 28466-U 28472-U 28471-U 28482-U 28564-U 28473-U
Colonnes capillaires, phases traditionnelles, SLB-5ms, Supelco®
Colonnes pour chromatographie
This column provides boiling point elution order with a slight increase in selectivity, especially for aromatic compounds.
  • Bonded and highly crosslinked
  • High thermal stability - maximum temperatures up to −60 to 330° C
  • Low bleed even at elevated temperatures