Imprimer
Sponge
Fournisseur: Nasco
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NASCB01351WAEA 510 EUR
NASCB01351WA
Sponge
This non sterile sponge can be used for a variety of surface sampling applications. The white, write-on strip makes sample identification easier and more convenient.
  • Sponge can be sterilised by radiation or autoclaving with the sponge placed in a glass or other autoclavable container
  • Users can easily write on the white strip using an ordinary ballpoint pen, pencil, or felt-tip marker
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904