Imprimer
Foetal bovine serum (FBS), EU origin
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S1780-100EA 102 EUR
BWSTS1780-100 BWSTS1820-500 BWSTS182H-100 BWSTS182H-500 BWSTS1780-500 BWSTS1820-100
Foetal bovine serum (FBS), EU origin
Sterile filtered foetal bovine serum (FBS) derived from clotted whole blood aseptically collected from foetus via cardiac puncture. Each manufactured batch is rigorously controlled, from the collection of serum through to final packaging.
  • EU origin
  • High quality serum, rigorously tested
  • BSE tested prior to production
  • Full traceability and security
Certifications: Biowest is a certified ISO 9001:2008 company. Registered by the French Ministry of Agriculture (EC regulation No. 1069/2009) under agreement no. FR 49.231.001 for the production of animal sera.