Imprimer
Biophotometers, BioSpectrometer® kinetic
Fournisseur: Eppendorf
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BioSpectrometer®
634-0812EA 0 EUR
634-0812 EPPE6136000.037 EPPE6136000.800
Biophotometers, BioSpectrometer® kinetic
Spectrophotomètres et Photomètres Biophotomètres
The BioSpectrometer kinetic combines the specifications of the basic model and a heating element.
  • Peltier element for temperature control of the shaft
  • Preprogrammed and freely programmable kinetic methods
  • Subsequent modification of the time window for regression evaluation of the kinetic curve
The cuvette shaft can be tempered in a range of 20 to 42 °C, which enables to carry out enzyme and substrate kinetics.