Imprimer
Botanical garden blot
Fournisseur: G-Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BB01EA 1570 EUR
BB01
Botanical garden blot
Réactifs d'électrophorèse Western Blotting Reagents
G-Biosciences' Botanical Garden Blot is prepared using total protein extracted from a variety of 5 to 10 day old, whole plant tissues.
Premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 - 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using Western transfer buffer. It is ready to be blocked and probed with the antibodies of choice.
Premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using Western transfer buffer. It is ready to be blocked and probed with the antibodies of choice.