Imprimer
RNAPure™, peqGOLD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
732-3312EA 162 EUR
PEQL30-1010
RNAPure™, peqGOLD
Ready to use reagent for quick and simple purification of high quality total RNA. Contains phenol and guanidine isothiocyanate.
  • Suitable for small and large samples
  • Reliable and reproducible with high purity RNA