Imprimer
DNA/RNA/protein purification reagent, peqGOLD TriFast™ and TriFast™ FL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
peqGOLD TriFast™
PEQL30-2010
DNA/RNA/protein purification reagent, peqGOLD TriFast™ and TriFast™ FL
For isolation of RNA, DNA and protein from one sample. Contains phenol and guanidine isothiocyanate.
  • Suitable for small and large samples
  • Specialised 'FL' version optimised for extractions from fluids