Imprimer
Presse à granulés manuelle, Port-A-Press™
Fournisseur: International Crystal Laboratories
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Port-A-Press
INCR0012-188EA 4650 EUR
INCR0012-188
Presse à granulés manuelle, Port-A-Press™
Presses
Port-A-Press™ is a full-scale KBr pellet press. It can produce pellets that are as transparent as those made with the 12 ton hydraulic press.
  • Very clear pellets
  • Faces of the anvils are optically polished
  • Scratch-free surface made of a hard nickel alloy
Uses a torque wrench to permit the user to apply the same amount of pressure from pellet to pellet thereby facilitating reproducibility of results.
Informations de commande: Complete with 7 mm die set, torque wrench, channel holder and pellet press.