Imprimer
Cloning discs, SP Bel-Art
Fournisseur: Bel-Art Products, a part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
391-2010EA 137 EUR
391-2010 391-2008 391-2009
Cloning discs, SP Bel-Art
Systèmes de stockage des acides nucléiques
These cloning discs quickly transfer cells to a 24-well plate. Saves time when isolating a large number of clones.
  • Free from RNase and DNase
  • Sterilised by gamma radiation
Cdt: Discs are packed in 2 vials, 50 discs per vial.