Imprimer
Anti-SRI Rabbit Polyclonal Antibody
Fournisseur: Aviva Systems Biology
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AVIVARP62987_P050EA 371 EUR
AVIVARP62987_P050
Anti-SRI Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: SRI (sorcin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant