Imprimer

Anti-SLC2A2 Goat Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA317322EA 666 EUR
ORIGTA317322
Anti-SLC2A2 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 2.5 µg/ml

Type: Primary
Antigen: SLC2A2 (solute carrier family 2 (facilitated glucose transporter), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR