Imprimer

Anti-SLC39A14 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA317329EA 666 EUR
ORIGTA317329
Anti-SLC39A14 Rabbit Polyclonal Antibody
Anticorps
  • Recommended Dilutions: Immunochemistry: 5-7.5 µg/ml

Type: Primary
Antigen: SLC39A14 (solute carrier family 39 (zinc transporter), member 14)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR