Imprimer

Anti-HRH3 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA317598EA 666 EUR
ORIGTA317598
Anti-HRH3 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 7-15 µg/ml

Type: Primary
Antigen: HRH3 (histamine receptor H3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR