Imprimer

Anti-PERP Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA317861EA 666 EUR
ORIGTA317861
Anti-PERP Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 5 µg/ml; Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: PERP (PERP, TP53 apoptosis effector)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR