Imprimer

Anti-APOH Goat Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA318083EA 666 EUR
ORIGTA318083
Anti-APOH Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA IE; Immunochemistry: 2.5 µg/ml

Type: Primary
Antigen: APOH (apolipoprotein H (beta-2-glycoprotein I))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR