Imprimer

Anti-CALB1 Mouse Monoclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA318675EA 666 EUR
ORIGTA318675
Anti-CALB1 Mouse Monoclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:100 Western Blot: 1:1000-1:2000

Type: Primary
Antigen: CALB1 (calbindin 1, 28kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR