Imprimer

Anti-SLITRK6 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA318847EA 666 EUR
ORIGTA318847
Anti-SLITRK6 Rabbit Polyclonal Antibody
Anticorps
  • Recommended Dilutions: Immunochemistry: 15 µg/ml

Type: Primary
Antigen: SLITRK6 (SLIT and NTRK-like family, member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR