Imprimer

Anti-BCL10 Mouse Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA400004EA 624 EUR
ORIGTA400004
Anti-BCL10 Mouse Antibody
Anticorps
Type: Primary
Antigen: BCL10 (B-cell CLL/lymphoma 10)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR