Imprimer

Anti-HES1 Mouse Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA400012EA 624 EUR
ORIGTA400012
Anti-HES1 Mouse Antibody
Anticorps
Type: Primary
Antigen: HES1 (hes family bHLH transcription factor 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR