Imprimer

Anti-KRT18 Mouse Monoclonal Antibody [clone: OTI1H10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500017EA 603 EUR
ORIGTA500017
Anti-KRT18 Mouse Monoclonal Antibody [clone: OTI1H10]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR