Imprimer

Anti-NOG Mouse Monoclonal Antibody [clone: OTI1H8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500029EA 608 EUR
ORIGTA500029
Anti-NOG Mouse Monoclonal Antibody [clone: OTI1H8]
Anticorps
Recommended Dilutions: Western Blot: 1:20000-40000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NOG (noggin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR