Imprimer

Anti-IL6 Mouse Monoclonal Antibody [clone: OTI2G3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500066EA 603 EUR
ORIGTA500066
Anti-IL6 Mouse Monoclonal Antibody [clone: OTI2G3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: IL6 (interleukin 6 (interferon, beta 2))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR