Imprimer

Anti-AKT3 Mouse Monoclonal Antibody [clone: OTI9H8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500080EA 603 EUR
ORIGTA500080
Anti-AKT3 Mouse Monoclonal Antibody [clone: OTI9H8]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1;50; Immunofluorescence: 1:100

Type: Primary
Antigen: AKT3 (v-akt murine thymoma viral oncogene homolog 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR