Imprimer

Anti-NEUROG3 Mouse Monoclonal Antibody [clone: OTI5A8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500090EA 603 EUR
ORIGTA500090
Anti-NEUROG3 Mouse Monoclonal Antibody [clone: OTI5A8]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: NEUROG3 (neurogenin 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR