Imprimer

Anti-SOX17 Mouse Monoclonal Antibody [clone: OTI2G8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500096EA 581 EUR
ORIGTA500096
Anti-SOX17 Mouse Monoclonal Antibody [clone: OTI2G8]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100

Type: Primary
Antigen: SOX17 (SRY (sex determining region Y)-box 17)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR