Imprimer

Anti-HHEX Mouse Monoclonal Antibody [clone: OTI3F4 1B1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500099EA 603 EUR
ORIGTA500099
Anti-HHEX Mouse Monoclonal Antibody [clone: OTI3F4 1B1]
Anticorps
Recommended Dilutions: Western Blot: 110000; Immunofluorescence: 1:100

Type: Primary
Antigen: HHEX (hematopoietically expressed homeobox)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR