Imprimer

Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI4F1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500170EA 581 EUR
ORIGTA500170
Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI4F1]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: CPA1 (carboxypeptidase A1 (pancreatic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR