Imprimer

Anti-KRT19 Mouse Monoclonal Antibody [clone: OTI3F8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA500212
ORIGTA500212EA 616 EUR
ORIGTA500212
Anti-KRT19 Mouse Monoclonal Antibody [clone: OTI3F8]
Anticorps
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: KRT19 (keratin 19)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR