Imprimer
Anti-NOTCH1 Mouse Monoclonal Antibody [clone: OTI4C9]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA500248EA 603 EUR
ORIGTA500248
Anti-NOTCH1 Mouse Monoclonal Antibody [clone: OTI4C9]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50
Type: Primary
Antigen: NOTCH1 (notch 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human