Imprimer

Anti-FGF2 Mouse Monoclonal Antibody [clone: OTI3D10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500269EA 603 EUR
ORIGTA500269
Anti-FGF2 Mouse Monoclonal Antibody [clone: OTI3D10]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: FGF2 (fibroblast growth factor 2 (basic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR