Imprimer

Anti-GAD1 Mouse Monoclonal Antibody [clone: OTI3H2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500328EA 603 EUR
ORIGTA500328
Anti-GAD1 Mouse Monoclonal Antibody [clone: OTI3H2]
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Immunoprecipitation: 4?g/ml

Type: Primary
Antigen: GAD1 (glutamate decarboxylase 1 (brain, 67kDa))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR