Imprimer
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: OTI11G2]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA500378EA 608 EUR
ORIGTA500378
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: OTI11G2]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunoprecipitation: 2?g/500?l
Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant