Imprimer

Anti-TYRO3 Mouse Monoclonal Antibody [clone: OTI5B7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500415EA 603 EUR
ORIGTA500415
Anti-TYRO3 Mouse Monoclonal Antibody [clone: OTI5B7]
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: TYRO3 (TYRO3 protein tyrosine kinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR