Imprimer

Anti-SSB Mouse Monoclonal Antibody [clone: OTI3F11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500436EA 603 EUR
ORIGTA500436
Anti-SSB Mouse Monoclonal Antibody [clone: OTI3F11]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SSB (Sjogren syndrome antigen B (autoantigen La))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR