Imprimer

Anti-BRAF Mouse Monoclonal Antibody [clone: OTI4B2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500438EA 603 EUR
ORIGTA500438
Anti-BRAF Mouse Monoclonal Antibody [clone: OTI4B2]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR