Imprimer

Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI4D10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500469EA 616 EUR
ORIGTA500469
Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI4D10]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Immunoprecipitation: 2-4μg/mg

Type: Primary
Antigen: MAPK1 (mitogen-activated protein kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR