Imprimer

Anti-CD4 Mouse Monoclonal Antibody [clone: OTI4F8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500488EA 581 EUR
ORIGTA500488
Anti-CD4 Mouse Monoclonal Antibody [clone: OTI4F8]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR