Imprimer
Anti-STK39 Mouse Monoclonal Antibody [clone: OTI4H3]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA500496EA 608 EUR
ORIGTA500496
Anti-STK39 Mouse Monoclonal Antibody [clone: OTI4H3]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:50; Flow Cytometry: 1:100
Type: Primary
Antigen: STK39 (serine threonine kinase 39)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human