Imprimer

Anti-STK39 Mouse Monoclonal Antibody [clone: OTI4E3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500501EA 603 EUR
ORIGTA500501
Anti-STK39 Mouse Monoclonal Antibody [clone: OTI4E3]
Anticorps
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: STK39 (serine threonine kinase 39)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR