Imprimer

Anti-TMEM100 Mouse Monoclonal Antibody [clone: OTI1F8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500508EA 608 EUR
ORIGTA500508
Anti-TMEM100 Mouse Monoclonal Antibody [clone: OTI1F8]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: TMEM100 (transmembrane protein 100)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR