Imprimer

Anti-BUB1B Mouse Monoclonal Antibody [clone: OTI11G2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500540EA 603 EUR
ORIGTA500540
Anti-BUB1B Mouse Monoclonal Antibody [clone: OTI11G2]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Immunofluorescence: 1:100

Type: Primary
Antigen: BUB1B (BUB1 mitotic checkpoint serine/threonine kinase B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR