Imprimer

Anti-PRKY Mouse Monoclonal Antibody [clone: OTI3D11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA500563EA 603 EUR
ORIGTA500563
Anti-PRKY Mouse Monoclonal Antibody [clone: OTI3D11]
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PRKY (protein kinase, Y-linked, pseudogene)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR